siRNA Details
Gene Symbol: ZNF410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242924.1 | NP_001229853.1 | 7 | 1194 | ||
NM_001242926.1 | NP_001229855.1 | 6 | 1143 | |||
NM_001242927.1 | NP_001229856.1 | 5 | 924 | |||
NM_001242928.1 | NP_001229857.1 | 6 | 1143 | |||
NM_021188.2 | NP_067011.1 | 6 | 1143 | |||
NR_040251.1 | 6 | 1111 | ||||
GenBank mRNA | AK223261.1 | - | 6 | 899 | ||
AK293350.1 | - | 7 | 950 | |||
AK298307.1 | - | 7 | 866 | |||
AK298520.1 | - | 6 | 919 | |||
AK299138.1 | - | 5 | 922 | |||
AK303834.1 | - | 6 | 809 | |||
AK304609.1 | - | 6 | 686 | |||
AK309957.1 | - | 7 | 939 | |||
AK310031.1 | - | 5 | 685 | |||
AK315206.1 | - | 6 | 901 | |||
BC004357.1 | - | 6 | 887 | |||
BC050683.1 | - | 6 | 953 | 5752452 | ||
U90919.1 | - | 6 | 875 |
