siRNA Details
Gene Symbol: ZNF350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021632.3 | NP_067645.3 | 5 | 893 | ||
XM_011527196.1 | XP_011525498.1 | 5 | 937 | |||
XM_017027094.1 | XP_016882583.1 | 6 | 1049 | |||
XM_017027095.1 | XP_016882584.1 | 6 | 1210 | |||
XM_017027096.1 | XP_016882585.1 | 6 | 1070 | |||
XM_017027097.1 | XP_016882586.1 | 6 | 1083 | |||
XM_017027098.1 | XP_016882587.1 | 6 | 1170 | |||
XM_017027099.1 | XP_016882588.1 | 7 | 1193 | |||
XM_017027100.1 | XP_016882589.1 | 5 | 1052 | |||
XM_017027101.1 | XP_016882590.1 | 6 | 1036 | |||
XM_017027102.1 | XP_016882591.1 | 6 | 1082 | |||
XM_017027103.1 | XP_016882592.1 | 6 | 1170 | |||
XM_017027104.1 | XP_016882593.1 | 6 | 1174 | |||
GenBank mRNA | AF295096.1 | - | 4 | 848 | ||
AF309561.1 | - | 5 | 882 | |||
AK023467.1 | - | 5 | 893 | |||
BC009921.2 | - | 4 | 833 | 3506774 |
