siRNA Details
Gene Symbol: SAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021818.3 | NP_068590.1 | 3 | 969 | ||
XM_011537057.2 | XP_011535359.1 | 4 | 1032 | |||
XM_017021568.1 | XP_016877057.1 | 4 | 1262 | |||
GenBank mRNA | AJ292969.1 | - | 3 | 748 | ||
AK021500.1 | - | 3 | 916 | |||
AK023071.1 | - | 3 | 798 | |||
AK095903.1 | - | 4 | 833 | |||
AK225896.1 | - | 3 | 941 | |||
AK290883.1 | - | 4 | 841 | |||
AL833378.1 | - | 2 | 379 | |||
BC020537.1 | - | 3 | 915 | 4425620 | ||
CR457297.1 | - | 3 | 583 |
