siRNA Details
Gene Symbol: WDR77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317062.1 | NP_001303991.1 | 2 | 422 | ||
NM_001317063.1 | NP_001303992.1 | 2 | 422 | |||
NM_001317064.1 | NP_001303993.1 | 2 | 422 | |||
NM_024102.3 | NP_077007.1 | 2 | 422 | |||
NR_133654.1 | 2 | 422 | ||||
GenBank mRNA | AB073603.1 | - | 2 | 370 | ||
AF052183.1 | - | 2 | 308 | |||
AF478464.1 | - | 2 | 283 | |||
AK022860.1 | - | 2 | 352 | |||
AK223189.1 | - | 2 | 352 | |||
AK296163.1 | - | 2 | 366 | |||
AK298179.1 | - | 2 | 366 | |||
AL557974.3 | - | 2 | 338 | |||
AY225316.1 | - | 2 | 308 | |||
BC001679.2 | - | 2 | 343 | 2822173 | ||
BC006477.1 | - | 2 | 343 | 2822173 | ||
BC009411.1 | - | 2 | 355 | 3614223 | ||
BC011778.2 | - | 2 | 362 | 3941878 | ||
BC016946.1 | - | 2 | 322 | 3845908 | ||
BI195029.1 | - | 1 | 94 | |||
DB105102.1 | - | 2 | 426 |
