siRNA Details
Gene Symbol: TMEM243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250589.3 | XP_005250646.1 | 5 | 687 | ||
XM_017012617.1 | XP_016868106.1 | 5 | 810 | |||
XM_017012618.1 | XP_016868107.1 | 5 | 554 | |||
GenBank mRNA | AK313794.1 | - | 4 | 489 | ||
BC002837.2 | - | 4 | 471 | 3634983 | ||
BT006973.1 | - | 4 | 259 | |||
DB241717.1 | - | 4 | 358 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081029, AK076719, BC147128, BC147127, BC147812, and others.
