siRNA Details
Gene Symbol: IRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024336.2 | NP_077312.2 | 2 | 1021 | ||
XM_005256139.3 | XP_005256196.1 | 2 | 1327 | |||
GenBank mRNA | AK309146.1 | - | 1 | 271 | ||
AY335943.1 | - | 2 | 1021 | |||
BC023667.2 | - | 2 | 988 | 5020235 |
