siRNA Details
Gene Symbol: IRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024336.2 | NP_077312.2 | 1 | 445 | ||
XM_005256139.3 | XP_005256196.1 | 1 | 751 | |||
GenBank mRNA | AY335943.1 | - | 1 | 445 | ||
BC023667.2 | - | 1 | 412 | 5020235 |
