siRNA Details
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024576.4 | NP_078852.3 | 2, 3 | 811 | ||
XM_017011304.1 | XP_016866793.1 | 2, 3 | 373 | |||
GenBank mRNA | BC018688.1 | - | 2, 3 | 446 | 4717736 | |
BC110999.1 | - | 2, 3 | 450 | 5585901 |
