siRNA Details
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324266.1 | NP_001311195.1 | 6 | 1161 | ||
NM_024576.4 | NP_078852.3 | 6 | 1158 | |||
XM_011536117.2 | XP_011534419.1 | 6 | 1094 | |||
XM_011536118.2 | XP_011534420.1 | 6 | 943 | |||
XM_017011303.1 | XP_016866792.1 | 6 | 723 | |||
XM_017011304.1 | XP_016866793.1 | 6 | 720 | |||
XM_017011305.1 | XP_016866794.1 | 6 | 616 | |||
XM_017011306.1 | XP_016866795.1 | 6 | 1646 | |||
GenBank mRNA | AK024732.1 | - | 3 | 221 | ||
BC018688.1 | - | 6 | 793 | 4717736 | ||
BC110999.1 | - | 6 | 797 | 5585901 |
