siRNA Details
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178055.1 | NP_001171526.1 | 25, 26 | 2557 | ||
NM_001178056.1 | NP_001171527.1 | 23, 24 | 2407 | |||
NM_024615.3 | NP_078891.2 | 24, 25 | 2533 | |||
XM_005248596.4 | XP_005248653.1 | 25, 26 | 2913 | |||
XM_011543631.2 | XP_011541933.1 | 25, 26 | 2446 | |||
XM_011543632.2 | XP_011541934.2 | 24, 25 | 2367 | |||
XM_011543634.2 | XP_011541936.1 | 26, 27 | 3042 | |||
XM_011543635.2 | XP_011541937.1 | 26, 27 | 2563 | |||
XM_011543636.2 | XP_011541938.1 | 25, 26 | 2427 | |||
XM_011543640.2 | XP_011541942.1 | 22, 23 | 2317 | |||
XM_011543641.2 | XP_011541943.1 | 23, 24 | 2294 | |||
XM_011543642.2 | XP_011541944.1 | 21, 22 | 2281 | |||
XM_011543643.2 | XP_011541945.1 | 22, 23 | 2722 | |||
XM_017009850.1 | XP_016865339.1 | 24, 25 | 2993 | |||
XM_017009851.1 | XP_016865340.1 | 23, 24 | 2433 | |||
GenBank mRNA | AK024961.1 | - | 14, 15 | 1603 | ||
AK225188.1 | - | 14, 15 | 1603 | |||
AK298680.1 | - | 23, 24 | 2396 | |||
AK314466.1 | - | 25, 26 | 2557 | |||
AL834477.1 | - | 24, 25 | 2492 | |||
BC075801.1 | - | 23, 24 | 2366 | 5226828 |
