siRNA Details
Gene Symbol: QTRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256835.1 | NP_001243764.1 | 4 | 469 | ||
NM_024638.3 | NP_078914.1 | 5 | 577 | |||
XM_005247769.3 | XP_005247826.1 | 5 | 358 | |||
XM_006713744.3 | XP_006713807.1 | 4 | 210 | |||
XM_011513137.2 | XP_011511439.1 | 4 | 440 | |||
GenBank mRNA | AK023022.1 | - | 5 | 467 | ||
AL833709.1 | - | 4 | 437 | |||
BC034559.2 | - | 4 | 451 | 4385906 | ||
BC039265.1 | - | 5 | 548 | 5296335 | ||
CR457296.1 | - | 3 | 318 | |||
DC400836.1 | - | 4 | 469 |
