siRNA Details
Gene Symbol: TBC1D17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168222.1 | NP_001161694.1 | 9 | 1298 | ||
NM_024682.2 | NP_078958.2 | 10 | 1397 | |||
XM_011527317.2 | XP_011525619.1 | 10 | 1309 | |||
GenBank mRNA | AB449903.1 | - | 10 | 1098 | ||
AK022230.1 | - | 10 | 1156 | |||
AK090606.1 | - | 9 | 1098 | |||
AK300007.1 | - | 9 | 1057 | |||
AY129013.1 | - | 8 | 1844 | |||
BC003516.1 | - | 10 | 1250 | 3050014 |
