siRNA Details
Gene Symbol: ZNF385D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024697.2 | NP_078973.1 | 5 | 962 | ||
XM_017007191.1 | XP_016862680.1 | 7 | 1000 | |||
XM_017007192.1 | XP_016862681.1 | 6 | 943 | |||
XM_017007193.1 | XP_016862682.1 | 7 | 990 | |||
XM_017007194.1 | XP_016862683.1 | 8 | 1055 | |||
XM_017007195.1 | XP_016862684.1 | 6 | 695 | |||
XM_017007196.1 | XP_016862685.1 | 6 | 1173 | |||
XM_017007197.1 | XP_016862686.1 | 6 | 1411 | |||
XM_017007198.1 | XP_016862687.1 | 6 | 1166 | |||
XM_017007199.1 | XP_016862688.1 | 5 | 521 | |||
XM_017007200.1 | XP_016862689.1 | 5 | 2737 | |||
XM_017007201.1 | XP_016862690.1 | 5 | 542 | |||
XM_017007202.1 | XP_016862691.1 | 3 | 258 | |||
GenBank mRNA | AK026072.1 | - | 5 | 962 | ||
AK310298.1 | - | 7 | 973 | |||
BC007212.1 | - | 5 | 786 | 3347783 |
