siRNA Details
Gene Symbol: ZNF665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024733.3 | NP_079009.3 | 3, 4 | 242 | ||
XM_005259266.4 | XP_005259323.1 | 3, 4 | 276 | |||
XM_011527325.2 | XP_011525627.1 | 3, 4 | 347 | |||
XM_011527327.2 | XP_011525629.1 | 4, 5 | 651 | |||
GenBank mRNA | AK291403.1 | - | 3, 4 | 232 | ||
AL832507.1 | - | 3, 4 | 227 |
