siRNA Details
Gene Symbol: AGMAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024758.4 | NP_079034.3 | 5 | 943 | ||
GenBank mRNA | AK130034.1 | - | 5 | 739 | ||
AK225366.1 | - | 5 | 856 | |||
AY057097.1 | - | 5 | 843 | |||
BC005090.1 | - | 5 | 943 | 3837251 |
