siRNA Details
Gene Symbol: RNF122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024787.3 | NP_079063.2 | 1, 2 | 422 | ||
GenBank mRNA | AK022588.1 | - | 1, 2 | 419 | ||
AU124463.1 | - | 1, 2 | 419 | |||
BC093884.1 | - | 1, 2 | 40 | 7939729 | ||
BC101573.1 | - | 1, 2 | 40 | 8069079 | ||
DA610129.1 | - | 1, 2 | 422 | |||
DA904646.1 | - | 1, 2 | 420 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175136, AK171823, BC038399, AK162034, AK136206, and others.
Gene Ontology Categories:
