siRNA Details
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031723.3 | NP_001026893.1 | 5 | 827 | ||
NM_001278310.1 | NP_001265239.1 | 4 | 655 | |||
XM_011532262.2 | XP_011530564.1 | 7 | 711 | |||
XM_011532263.2 | XP_011530565.1 | 5 | 1211 | |||
XM_011532265.2 | XP_011530567.1 | 4 | 1155 | |||
XM_017008628.1 | XP_016864117.1 | 7 | 893 | |||
GenBank mRNA | AK024343.1 | - | 3 | 354 | ||
AL833076.2 | - | 4 | 2699 | |||
AL833412.1 | - | 4 | 517 | |||
AY186740.1 | - | 7 | 706 | |||
AY358272.1 | - | 5 | 681 | |||
BC022248.1 | - | 5 | 696 | 4771526 |
Gene Symbol: LOC646358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107769.1 | - | 1 | 57 |
