siRNA Details
Gene Symbol: ANKRD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115116.1 | NP_001108588.1 | 4 | 936 | ||
NM_024933.3 | NP_079209.3 | 4 | 936 | |||
XM_005264569.4 | XP_005264626.1 | 4 | 2096 | |||
GenBank mRNA | AK093479.1 | - | 4 | 772 | ||
BC035234.1 | - | 4 | 936 | 5166484 | ||
DB036445.1 | - | 3 | 534 |
