siRNA Details
Gene Symbol: RMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024945.2 | NP_079221.2 | 3 | 832 | ||
XM_005252211.2 | XP_005252268.1 | 3 | 701 | |||
XM_005252213.2 | XP_005252270.1 | 4 | 777 | |||
XM_011519034.2 | XP_011517336.1 | 4 | 890 | |||
XM_017015140.1 | XP_016870629.1 | 3 | 642 | |||
GenBank mRNA | AK022950.1 | - | 3 | 756 | ||
BC020606.1 | - | 3 | 654 | |||
BC032494.1 | - | 4 | 768 | |||
BC039999.2 | - | 3 | 659 | 5554889 | ||
BC053549.1 | - | 3 | 592 | |||
BC064937.1 | - | 2 | 572 | 6158300 | ||
DN997105.1 | - | 3 | 704 |
