siRNA Details
Gene Symbol: ZNF430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172671.1 | NP_001166142.1 | 1, 2 | 194 | ||
NM_025189.3 | NP_079465.3 | 1, 2 | 194 | |||
GenBank mRNA | AK023721.1 | - | 1, 2 | 174 | ||
AK290475.1 | - | 1, 2 | 194 | |||
AY269787.1 | - | 1, 2 | 123 | |||
BC111776.1 | - | 1, 2 | 82 | 40033720 | ||
DB255551.1 | - | 1, 2 | 157 |
