siRNA Details
Gene Symbol: TTYH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025250.2 | NP_079526.1 | 12, 13 | 1606 | ||
XM_011515535.2 | XP_011513837.1 | 12, 13 | 1574 | |||
XM_017012655.1 | XP_016868144.1 | 11, 12 | 1480 | |||
XM_017012656.1 | XP_016868145.1 | 11, 12 | 1478 | |||
GenBank mRNA | AB051478.2 | - | 12, 13 | 1601 | ||
AB162931.1 | - | 12, 13 | 1431 | |||
AK074158.1 | - | 8, 9 | 914 | |||
AK124608.1 | - | 8, 9 | 1049 | |||
BC131824.1 | - | 12, 13 | 1477 | 40128913 | ||
BC152447.1 | - | 12, 13 | 1601 | 8862578 |
