siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 4820 | |||
GenBank mRNA | AF251293.1 | - | 1 | 509 | ||
AK025814.1 | - | 1 | 1982 | |||
AK123844.1 | - | 1 | 2550 | |||
BC015678.2 | - | 1 | 2810 | |||
BC034918.1 | - | 1 | 772 | |||
BX648949.1 | - | 1 | 4467 | |||
CR749230.1 | - | 1 | 4825 |
