siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 4952 | |||
GenBank mRNA | AF251293.1 | - | 1 | 641 | ||
AJ420426.1 | - | 1 | 123 | |||
AK025814.1 | - | 1 | 2114 | |||
AK123844.1 | - | 1 | 2682 | |||
BC015678.2 | - | 1 | 2942 | |||
BC034918.1 | - | 1 | 904 | |||
CR749230.1 | - | 1 | 4957 |
