siRNA Details
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 5 | 812 | ||
NM_001160224.1 | NP_001153696.1 | 5 | 582 | |||
NM_001160225.1 | NP_001153697.1 | 5 | 461 | |||
NM_030954.3 | NP_112216.3 | 5 | 582 | |||
XM_006716404.2 | XP_006716467.1 | 6 | 569 | |||
XM_011544666.2 | XP_011542968.1 | 5 | 366 | |||
XM_017013881.1 | XP_016869370.1 | 6 | 509 | |||
XM_017013882.1 | XP_016869371.1 | 4 | 354 | |||
GenBank mRNA | AK027748.1 | - | 5 | 498 | ||
AK225086.1 | - | 5 | 462 | |||
AK225568.1 | - | 5 | 478 | |||
AL136620.1 | - | 5 | 466 | |||
BC032393.1 | - | 5 | 444 | 5200847 | ||
BC039461.1 | - | 5 | 569 | |||
BC044566.1 | - | 5 | 582 | |||
BC058289.1 | - | 5 | 445 |
