siRNA Details
Gene Symbol: C19orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031726.3 | NP_001026896.2 | 3 | 365 | ||
NM_001256046.1 | NP_001242975.1 | 3 | 636 | |||
NM_001256047.1 | NP_001242976.1 | 3 | 442 | |||
NM_001282929.1 | NP_001269858.1 | 2 | 195 | |||
NM_001282930.1 | NP_001269859.1 | 2 | 466 | |||
NM_001282931.1 | NP_001269860.1 | 4 | 757 | |||
NM_031448.4 | NP_113636.2 | 3 | 636 | |||
GenBank mRNA | AK057185.1 | - | 3 | 333 | ||
AL162066.1 | - | 2 | 140 | |||
BC004957.1 | - | 2 | 126 | 3628073 | ||
BC009946.2 | - | 2 | 126 | 4125477 | ||
BC017211.2 | - | 1 | 26 | |||
BC063518.1 | - | 3 | 315 | 4340755 | ||
BG720088.1 | - | 4 | 485 | |||
BG722048.1 | - | 3 | 604 | |||
BI753584.1 | - | 3 | 368 | |||
DA064969.1 | - | 2 | 466 | |||
DA189248.1 | - | 3 | 442 | |||
DA348401.1 | - | 2 | 187 | |||
DA708831.1 | - | 3 | 358 |
