siRNA Details
Gene Symbol: C19orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031726.3 | NP_001026896.2 | 3 | 571 | ||
NM_001256046.1 | NP_001242975.1 | 4 | 787 | |||
NM_001256047.1 | NP_001242976.1 | 3 | 648 | |||
NM_001282929.1 | NP_001269858.1 | 2 | 401 | |||
NM_001282930.1 | NP_001269859.1 | 2 | 672 | |||
NM_001282931.1 | NP_001269860.1 | 4 | 963 | |||
NM_031448.4 | NP_113636.2 | 3 | 842 | |||
GenBank mRNA | AK057185.1 | - | 3 | 539 | ||
AL162066.1 | - | 2 | 346 | |||
BC004957.1 | - | 2 | 332 | 3628073 | ||
BC009946.2 | - | 2 | 332 | 4125477 | ||
BC017211.2 | - | 1 | 232 | |||
BC063518.1 | - | 4 | 466 | 4340755 | ||
BI753584.1 | - | 3 | 574 | |||
DA348401.1 | - | 2 | 393 |
