siRNA Details
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 11, 12 | 1645 | ||
NM_031449.3 | NP_113637.3 | 12, 13 | 1814 | |||
NM_174929.2 | NP_777589.2 | 10, 11 | 1675 | |||
XM_005249866.2 | XP_005249923.1 | 12, 13 | 1865 | |||
XM_005249867.4 | XP_005249924.1 | 12, 13 | 1867 | |||
XM_005249868.4 | XP_005249925.1 | 12, 13 | 2885 | |||
XM_005249869.2 | XP_005249926.1 | 12, 13 | 1838 | |||
XM_005249870.2 | XP_005249927.1 | 11, 12 | 1787 | |||
XM_005249871.2 | XP_005249928.1 | 12, 13 | 1769 | |||
XM_005249872.2 | XP_005249929.1 | 11, 12 | 1760 | |||
XM_005249873.2 | XP_005249930.1 | 12, 13 | 1742 | |||
XM_011515566.1 | XP_011513868.1 | 6, 7 | 1005 | |||
XM_017012674.1 | XP_016868163.1 | 12, 13 | 1946 | |||
GenBank mRNA | AB067473.1 | - | 6, 7 | 898 | ||
AF357908.1 | - | 11, 12 | 1697 | |||
AK090415.1 | - | 12, 13 | 1814 | |||
AK090841.1 | - | 4, 5 | 1094 | |||
AL136572.1 | - | 2, 3 | 267 | |||
AL831933.1 | - | 3, 4 | 1412 | |||
AY426594.1 | - | 10, 11 | 1637 | |||
BC016968.1 | - | 10, 11 | 1658 | |||
BC021924.1 | - | 6, 7 | 746 | |||
BC110435.1 | - | 11, 12 | 1630 | 5518312 |

More Information
Other Species Targeted

Gene Ontology Categories:
