siRNA Details
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB067469.1 | - | 19, 20 | 2893 | ||
AK026394.1 | - | 19, 20 | 2935 | |||
AK027689.1 | - | 8, 9 | 1043 | |||
AK128755.1 | - | 19, 20 | 3119 | |||
AK308815.1 | - | 8, 9 | 1214 | |||
AL833973.1 | - | 17, 18 | 2218 | |||
AY190606.1 | - | 17, 18 | 2197 | |||
BC003650.2 | - | 5, 6 | 662 | |||
BC006206.2 | - | 17, 18 | 2339 | 3538806 | ||
BC065288.1 | - | 2, 3 | 203 |
