siRNA Details
Gene Symbol: MON1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142501.1 | NP_001135973.1 | 3 | 1268 | ||
NM_032355.3 | NP_115731.2 | 4 | 1754 | |||
XM_006713345.3 | XP_006713408.1 | 4 | 1520 | |||
XM_011534160.1 | XP_011532462.1 | 4 | 1383 | |||
GenBank mRNA | AK074404.1 | - | 4 | 1564 | ||
AK092018.1 | - | 3 | 1067 | |||
AK315630.1 | - | 4 | 1608 | |||
BC006299.2 | - | 4 | 1566 | 4079816 | ||
BC009459.1 | - | 4 | 1523 | 3536968 | ||
BC047022.1 | - | 3 | 1018 | 5248424 |
