siRNA Details
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300912.1 | NP_001287841.1 | 5 | 412 | ||
NM_032433.2 | NP_115809.1 | 5 | 394 | |||
XM_005260098.3 | XP_005260155.1 | 5 | 403 | |||
XM_006722924.2 | XP_006722987.1 | 5 | 412 | |||
XM_011528362.2 | XP_011526664.1 | 5 | 435 | |||
XM_011528363.2 | XP_011526665.1 | 5 | 353 | |||
XM_011528365.2 | XP_011526667.1 | 5 | 353 | |||
XM_011528366.2 | XP_011526668.1 | 5 | 403 | |||
XM_011528367.2 | XP_011526669.1 | 5 | 403 | |||
XM_011528368.2 | XP_011526670.1 | 5 | 323 | |||
XM_011528369.2 | XP_011526671.1 | 5 | 323 | |||
XM_017027367.1 | XP_016882856.1 | 5 | 411 | |||
XM_017027368.1 | XP_016882857.1 | 4 | 279 | |||
XM_017027369.1 | XP_016882858.1 | 5 | 353 | |||
XM_017027370.1 | XP_016882859.1 | 5 | 353 | |||
XM_017027371.1 | XP_016882860.1 | 5 | 403 | |||
GenBank mRNA | AF372702.1 | - | 5 | 351 | ||
AK094028.1 | - | 5 | 323 | |||
AK291471.1 | - | 5 | 421 | |||
AK307147.1 | - | 5 | 404 | |||
AK309208.1 | - | 5 | 346 | |||
BC048107.1 | - | 5 | 383 | |||
BC064571.1 | - | 5 | 394 | |||
BX647475.1 | - | 5 | 355 | |||
DA103403.1 | - | 5 | 412 |
