siRNA Details
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 14 | 1606 | ||
NM_032830.2 | NP_116219.2 | 14 | 1660 | |||
XM_005256205.3 | XP_005256262.2 | 10 | 1191 | |||
GenBank mRNA | AB075868.2 | - | 14 | 1598 | ||
AK027419.1 | - | 11 | 1215 | |||
AK027445.1 | - | 9 | 938 | |||
AK027584.1 | - | 3 | 237 | |||
AK027634.1 | - | 14 | 1612 | |||
AK027675.1 | - | 14 | 1609 | |||
AK074795.1 | - | 14 | 1626 | |||
BC000167.2 | - | 10 | 1105 | |||
BC009348.2 | - | 14 | 1592 | 4131625 | ||
BX647265.1 | - | 14 | 1574 | |||
CA430627.1 | - | 2 | 308 |
