siRNA Details
Gene Symbol: TNKS1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033396.2 | NP_203754.2 | 6 | 3025 | ||
XM_006718725.3 | XP_006718788.1 | 6 | 3041 | |||
XM_011545325.2 | XP_011543627.1 | 6 | 3038 | |||
GenBank mRNA | AB051528.2 | - | 6 | 3005 | ||
AF441771.1 | - | 5 | 3167 | |||
AK074113.1 | - | 2 | 1380 | |||
AK124903.1 | - | 6 | 3052 | |||
AK127871.1 | - | 4 | 2448 | |||
BC150333.1 | - | 6 | 3016 | 8860508 |
