siRNA Details
Gene Symbol: OTULIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138348.4 | NP_612357.4 | 7 | 1082 | ||
XM_011514151.2 | XP_011512453.1 | 7 | 4962 | |||
XM_011514152.2 | XP_011512454.1 | 7 | 4962 | |||
XM_011514154.2 | XP_011512456.1 | 6 | 4692 | |||
XM_017010015.1 | XP_016865504.1 | 6 | 4885 | |||
GenBank mRNA | AK092203.1 | - | 3 | 402 | ||
BC007706.2 | - | 5 | 655 | 3952677 | ||
BC015392.1 | - | 7 | 1008 | 4430188 |
