siRNA Details
Gene Symbol: C1orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139240.3 | NP_640333.3 | 2 | 271 | ||
XM_005245608.2 | XP_005245665.1 | 2 | 135 | |||
XM_011510152.2 | XP_011508454.1 | 4 | 2480 | |||
XM_011510153.2 | XP_011508455.1 | 3 | 2443 | |||
XM_011510155.2 | XP_011508457.1 | 4 | 2480 | |||
XM_017002808.1 | XP_016858297.1 | 2 | 69 | |||
GenBank mRNA | AL035295.1 | - | 2 | 271 | ||
BC038410.1 | - | 2 | 269 | 5169137 |
