siRNA Details
Gene Symbol: C1orf158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152290.3 | NP_689503.3 | 1 | 306 | ||
XM_011542434.1 | XP_011540736.1 | 1 | 291 | |||
GenBank mRNA | AK298760.1 | - | 1 | 268 | ||
BC029894.1 | - | 1 | 277 | 5171302 | ||
HY002971.1 | - | 1 | 307 |
