siRNA Details
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001552.4 | NP_001001552.3 | 4 | 421 | ||
NM_001199050.1 | NP_001185979.1 | 6 | 563 | |||
NM_001199051.1 | NP_001185980.1 | 5 | 440 | |||
NM_001199052.1 | NP_001185981.1 | 3 | 298 | |||
NR_037583.1 | 4 | 375 | ||||
XM_011510160.2 | XP_011508462.1 | 7 | 651 | |||
XM_011510161.2 | XP_011508463.1 | 7 | 639 | |||
XM_011510162.2 | XP_011508464.1 | 7 | 631 | |||
XM_011510163.2 | XP_011508465.1 | 6 | 531 | |||
XM_011510164.2 | XP_011508466.1 | 8 | 718 | |||
XM_011510165.2 | XP_011508467.1 | 8 | 690 | |||
GenBank mRNA | AB084764.1 | - | 3 | 265 | ||
AB084765.1 | - | 4 | 342 | |||
AB085610.1 | - | 4 | 388 | |||
AB096677.1 | - | 6 | 530 | |||
AB096678.1 | - | 4 | 342 | |||
AB096679.1 | - | 5 | 407 | |||
AB096680.1 | - | 5 | 465 | |||
AB096681.1 | - | 4 | 388 | |||
AB096682.1 | - | 3 | 265 | |||
AI923802.1 | - | 1 | 401 | |||
AK026628.1 | - | 4 | 388 | |||
AK302392.1 | - | 6 | 565 | |||
BC036636.2 | - | 6 | 533 | 5296437 |
