siRNA Details
Gene Symbol: DIRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052952.2 | NP_443184.1 | 2 | 336 | ||
XM_017003281.1 | XP_016858770.1 | 2 | 157 | |||
GenBank mRNA | AY039011.1 | - | 2 | 337 | ||
BC125137.1 | - | 1 | 93 | 40119557 |
