siRNA Details
Gene Symbol: SLC18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052831.2 | NP_439896.1 | 8 | 967 | ||
XM_017010236.1 | XP_016865725.1 | 8 | 1158 | |||
GenBank mRNA | AJ420537.1 | - | 2 | 93 | ||
AK124442.1 | - | 8 | 846 | |||
AK289918.1 | - | 8 | 986 | |||
AK293853.1 | - | 7 | 773 | |||
BC069567.1 | - | 4 | 370 | |||
BC069570.1 | - | 8 | 887 | 7262298 |
