siRNA Details
Gene Symbol: OR4X2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004727.1 | NP_001004727.1 | 1 | 561 | ||
GenBank mRNA | BC136935.1 | - | 1 | 609 | 9020927 | |
BC136936.1 | - | 1 | 609 | 9020928 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001270423, NM_146263, NM_146265, CB174322, BC145937.
Gene Ontology Categories:
