siRNA Details
Gene Symbol: SPACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145248.4 | NP_660291.2 | 4 | 330 | ||
XM_011537466.1 | XP_011535768.1 | 4 | 355 | |||
XM_011537467.2 | XP_011535769.1 | 3 | 298 | |||
XM_011537468.2 | XP_011535770.1 | 5 | 416 | |||
XM_011537471.2 | XP_011535773.1 | 5 | 486 | |||
GenBank mRNA | AI219896.1 | - | 4 | 453 | ||
AK130866.1 | - | 4 | 330 | |||
BC016750.2 | - | 4 | 312 | 4837062 |
