siRNA Details
Gene Symbol: SPACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145248.4 | NP_660291.2 | 5 | 431 | ||
XM_011537466.1 | XP_011535768.1 | 5 | 456 | |||
XM_011537467.2 | XP_011535769.1 | 4 | 399 | |||
XM_011537468.2 | XP_011535770.1 | 6 | 517 | |||
XM_011537471.2 | XP_011535773.1 | 6 | 587 | |||
XM_011537472.2 | XP_011535774.1 | 3 | 268 | |||
XM_017020391.1 | XP_016875880.1 | 2 | 213 | |||
XM_017020392.1 | XP_016875881.1 | 4 | 334 | |||
GenBank mRNA | AI219896.1 | - | 3 | 352 | ||
AK130866.1 | - | 5 | 431 | |||
BC016750.2 | - | 5 | 413 | 4837062 |
