siRNA Details
Gene Symbol: NOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113475.2 | NP_001106946.1 | 3, 4 | 695 | ||
XM_005267330.3 | XP_005267387.1 | 4, 5 | 1320 | |||
XM_011536427.2 | XP_011534729.1 | 4, 5 | 1333 | |||
XM_011536428.2 | XP_011534730.1 | 4, 5 | 580 | |||
XM_011536429.2 | XP_011534731.1 | 2, 3 | 608 | |||
GenBank mRNA | AK057371.1 | - | 3, 4 | 696 | ||
BC066965.1 | - | 3, 4 | 675 | 4373886 | ||
BI829015.1 | - | 4, 5 | 587 |
