siRNA Details
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 4 | 449 | ||
GenBank mRNA | AK128671.1 | - | 2 | 195 | ||
AK299345.1 | - | 5 | 526 | |||
AL832197.1 | - | 4 | 446 | |||
BC018134.1 | - | 4 | 404 | 3912539 | ||
BC055408.1 | - | 4 | 444 | 3684126 |
