siRNA Details
Gene Symbol: KRBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304947.1 | NP_001291876.1 | 2 | 1971 | ||
NM_213597.3 | NP_998762.1 | 2 | 1391 | |||
XM_005256443.2 | XP_005256500.1 | 5 | 2548 | |||
XM_011523643.1 | XP_011521945.1 | 4 | 2364 | |||
XM_011523644.1 | XP_011521946.1 | 3 | 2155 | |||
XM_017024163.1 | XP_016879652.1 | 4 | 2637 | |||
XM_017024164.1 | XP_016879653.1 | 2 | 1626 | |||
XM_017024165.1 | XP_016879654.1 | 3 | 2451 | |||
GenBank mRNA | AK131219.1 | - | 2 | 1392 | ||
BC033390.1 | - | 2 | 1226 |
