siRNA Details
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289002.1 | NP_001275931.1 | 3 | 957 | ||
NM_173488.4 | NP_775759.3 | 3 | 957 | |||
XM_005271874.3 | XP_005271931.1 | 3 | 4154 | |||
XM_011543147.2 | XP_011541449.1 | 3 | 4049 | |||
XM_011543148.2 | XP_011541450.1 | 3 | 3917 | |||
XM_011543150.1 | XP_011541452.1 | 2 | 167 | |||
XM_011543154.2 | XP_011541456.1 | 3 | 4154 | |||
GenBank mRNA | AF505657.1 | - | 3 | 928 | ||
AY273897.1 | - | 3 | 920 |
