siRNA Details
Gene Symbol: ZNF572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152412.2 | NP_689625.2 | 3 | 390 | ||
GenBank mRNA | AK095321.1 | - | 3 | 377 | ||
BC130515.1 | - | 3 | 369 | 40146546 | ||
BC130517.1 | - | 3 | 369 | 40146548 | ||
BX537876.1 | - | 3 | 390 | |||
DB093007.1 | - | 3 | 390 |
