siRNA Details
Gene Symbol: TCEAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080390.3 | NP_525129.1 | 3 | 663 | ||
GenBank mRNA | AF325115.1 | - | 3 | 608 | ||
AK312138.1 | - | 3 | 636 | |||
BC093932.1 | - | 2 | 541 | 7939777 | ||
BC104764.1 | - | 2 | 542 | 8143767 | ||
BM699243.1 | - | 1 | 39 |
