siRNA Details
Gene Symbol: SPATA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173499.4 | NP_775770.1 | 1 | 290 | ||
XM_011521265.2 | XP_011519567.1 | 1 | 251 | |||
GenBank mRNA | AY489187.1 | - | 1 | 254 | ||
BC033979.2 | - | 1 | 227 | 5295377 | ||
BC066292.1 | - | 1 | 249 | 5271997 | ||
BC112390.1 | - | 1 | 210 | 5168326 |
