siRNA Details
Gene Symbol: IGFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002915.2 | NP_001002915.2 | 2, 3 | 339 | ||
XM_011526501.1 | XP_011524803.1 | 2, 3 | 2878 | |||
XM_017026335.1 | XP_016881824.1 | 1, 2 | 727 | |||
GenBank mRNA | AY672112.1 | - | 2, 3 | 339 | ||
BC130490.1 | - | 1, 2 | 140 | 40146521 | ||
BC130492.1 | - | 1, 2 | 140 | 40146523 |
