siRNA Details
Gene Symbol: ZNF385B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113397.1 | NP_001106868.1 | 8 | 1478 | ||
NM_001113398.1 | NP_001106869.1 | 8 | 1338 | |||
NM_001282725.1 | NP_001269654.1 | 10 | 1432 | |||
NM_152520.4 | NP_689733.3 | 10 | 2005 | |||
NR_104234.1 | 12 | 1665 | ||||
XM_011510713.2 | XP_011509015.1 | 12 | 5146 | |||
XM_011510714.2 | XP_011509016.1 | 11 | 5096 | |||
XM_011510715.2 | XP_011509017.1 | 11 | 5049 | |||
XM_011510716.2 | XP_011509018.1 | 11 | 2257 | |||
XM_011510717.2 | XP_011509019.1 | 11 | 2254 | |||
XM_011510719.2 | XP_011509021.1 | 11 | 1517 | |||
XM_011510720.2 | XP_011509022.1 | 10 | 1377 | |||
XM_011510721.2 | XP_011509023.1 | 9 | 3670 | |||
XM_017003435.1 | XP_016858924.1 | 10 | 1686 | |||
XM_017003436.1 | XP_016858925.1 | 10 | 2164 | |||
XM_017003437.1 | XP_016858926.1 | 9 | 1321 | |||
XM_017003438.1 | XP_016858927.1 | 8 | 6804 | |||
XM_017003439.1 | XP_016858928.1 | 8 | 1312 | |||
XM_017003440.1 | XP_016858929.1 | 9 | 4841 | |||
GenBank mRNA | AK057999.1 | - | 8 | 1338 | ||
AK096822.1 | - | 8 | 1478 | |||
BC038422.1 | - | 10 | 1430 | 5165975 | ||
BC048123.1 | - | 12 | 1668 | |||
BC092423.1 | - | 10 | 1840 | 30378128 |
